Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAAATGCACAAAACTTAGCTAGC[A/C]CTAAAAGAAATACTGAATTTTCCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614649 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF170 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RNF170 - ring finger protein 170 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001160223.1 | 1275 | UTR 3 | NP_001153695.1 | |||
NM_001160224.1 | 1275 | Intron | NP_001153696.1 | |||
NM_001160225.1 | 1275 | UTR 3 | NP_001153697.1 | |||
NM_030954.3 | 1275 | UTR 3 | NP_112216.3 | |||
XM_006716404.2 | 1275 | UTR 3 | XP_006716467.1 | |||
XM_006716405.2 | 1275 | UTR 3 | XP_006716468.1 | |||
XM_011544666.2 | 1275 | UTR 3 | XP_011542968.1 | |||
XM_017013881.1 | 1275 | UTR 3 | XP_016869370.1 | |||
XM_017013882.1 | 1275 | UTR 3 | XP_016869371.1 |