Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACGTTGAACTCAGTGTATACTTG[G/T]GAAAATAACCTGCCATTTATTTTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601385 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TUSC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TUSC3 - tumor suppressor candidate 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006765.3 | Intron | NP_006756.2 | ||||
NM_178234.2 | Intron | NP_839952.1 | ||||
XM_011544651.2 | Intron | XP_011542953.1 | ||||
XM_011544652.2 | Intron | XP_011542954.1 | ||||
XM_011544654.2 | Intron | XP_011542956.1 | ||||
XM_017013860.1 | Intron | XP_016869349.1 | ||||
XM_017013861.1 | Intron | XP_016869350.1 |