Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTCGCGGGGTACCCCATCTTCCC[C/T]GGGACTGCCTTGATAATCCCCTGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114220 MIM: 611696 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPN1 - calpain 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A20 - solute carrier family 22 member 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004326.4 | Intron | NP_001004326.4 | ||||
XM_011545043.2 | Intron | XP_011543345.1 | ||||
XM_017017774.1 | Intron | XP_016873263.1 | ||||
XM_017017775.1 | Intron | XP_016873264.1 | ||||
XM_017017776.1 | Intron | XP_016873265.1 |