Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAGGAGGCCAGCACCACTCGGCGG[G/T]TCTTCTGGATCAAAGGGTGTTGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608965 | ||||||||||||||||||||
Literature Links: |
CABP4 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CABP4 - calcium binding protein 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM134 - transmembrane protein 134 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001078650.2 | 444 | Missense Mutation | AAC,ACC | N,T 120 | NP_001072118.1 | |
NM_001078651.2 | 444 | Missense Mutation | AAC,ACC | N,T 111 | NP_001072119.1 | |
NM_025124.3 | 444 | Missense Mutation | AAC,ACC | N,T 120 | NP_079400.1 | |
XM_006718693.2 | 444 | Missense Mutation | AAC,ACC | N,T 111 | XP_006718756.1 | |
XM_011545268.2 | 444 | Missense Mutation | AAC,ACC | N,T 120 | XP_011543570.1 | |
XM_017018356.1 | 444 | Missense Mutation | AAC,ACC | N,T 120 | XP_016873845.1 |