Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTACCCATTATTTTTAGTTCAT[C/T]TTCCAAGTGCTAAAGAGTCTGAAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600528 MIM: 604374 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CPT1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CPT1A - carnitine palmitoyltransferase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031847.2 | Intron | NP_001027017.1 | ||||
NM_001876.3 | Intron | NP_001867.2 | ||||
XM_005273762.2 | Intron | XP_005273819.1 | ||||
XM_005273763.1 | Intron | XP_005273820.1 | ||||
XM_017017220.1 | Intron | XP_016872709.1 |
TESMIN - testis expressed metallothionein like protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |