Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCCATCGGCAGAGCGCATGCGT[A/G]GCACAGCTACTACAATGAGAGTGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR5M3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR5M3 - olfactory receptor family 5 subfamily M member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR5M9 - olfactory receptor family 5 subfamily M member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004743.1 | 671 | Missense Mutation | CCA,CTA | P,L 224 | NP_001004743.1 |