Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGGAACACAGACCCAACAAGA[A/T]TTTATTTATACAAGAAAAGGGACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600454 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
KLHL35 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KLHL35 - kelch like family member 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039548.2 | 1881 | UTR 3 | NP_001034637.2 | |||
XM_011544954.2 | 1881 | UTR 3 | XP_011543256.1 |
RPS3 - ribosomal protein S3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005.4 | 1881 | Intron | NP_000996.2 | |||
NM_001256802.1 | 1881 | Intron | NP_001243731.1 | |||
NM_001260506.1 | 1881 | Intron | NP_001247435.1 | |||
NM_001260507.1 | 1881 | Intron | NP_001247436.1 |