Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTACCTGTGGTGATTAGTTTTAT[C/T]TGTGAACTCGGCTAGGCTATAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 617036 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACER3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ACER3 - alkaline ceramidase 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300953.1 | Intron | NP_001287882.1 | ||||
NM_001300954.1 | Intron | NP_001287883.1 | ||||
NM_001300955.1 | Intron | NP_001287884.1 | ||||
NM_018367.6 | Intron | NP_060837.3 | ||||
XM_011545151.2 | Intron | XP_011543453.1 | ||||
XM_011545152.2 | Intron | XP_011543454.1 | ||||
XM_011545153.2 | Intron | XP_011543455.1 | ||||
XM_017017987.1 | Intron | XP_016873476.1 | ||||
XM_017017988.1 | Intron | XP_016873477.1 |