Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATGCAGAAGCCATCGCTCAGAGC[A/G]ATTTTCAGTACCTCTTCCACGAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
KCTD21 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
KCTD21 - potassium channel tetramerization domain containing 21 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001029859.2 | 776 | Silent Mutation | ATC,ATT | I,I 230 | NP_001025030.1 | |
XM_005273925.3 | 776 | Silent Mutation | ATC,ATT | I,I 299 | XP_005273982.1 | |
XM_005273928.1 | 776 | Silent Mutation | ATC,ATT | I,I 230 | XP_005273985.1 | |
XM_006718516.3 | 776 | Silent Mutation | ATC,ATT | I,I 266 | XP_006718579.1 | |
XM_006718517.2 | 776 | Silent Mutation | ATC,ATT | I,I 230 | XP_006718580.1 | |
XM_006718518.3 | 776 | Silent Mutation | ATC,ATT | I,I 230 | XP_006718581.1 | |
XM_011544955.2 | 776 | Silent Mutation | ATC,ATT | I,I 230 | XP_011543257.1 |
KCTD21-AS1 - KCTD21 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |