Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGCGGACTCCTTCTGGCTCGACTT[A/G]CTCTCAGCCAGCTCTCCCTCCTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607104 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNKS1BP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNKS1BP1 - tankyrase 1 binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033396.2 | 5190 | Silent Mutation | AGC,AGT | S,S 1675 | NP_203754.2 | |
XM_006718725.3 | 5190 | Silent Mutation | AGC,AGT | S,S 1675 | XP_006718788.1 | |
XM_011545325.2 | 5190 | Intron | XP_011543627.1 |