Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTCACAGATTCCCCCTATGGCAG[G/T]TAAAGCTCCTCATCAGGTTCCTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608876 | ||||||||||||||||||||
Literature Links: |
PCF11 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCF11 - PCF11 cleavage and polyadenylation factor subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015885.3 | 1142 | Missense Mutation | GGT,GTT | G,V 266 | NP_056969.2 | |
XM_005274048.1 | 1142 | Missense Mutation | GGT,GTT | G,V 266 | XP_005274105.1 | |
XM_005274049.1 | 1142 | Missense Mutation | GGT,GTT | G,V 266 | XP_005274106.1 | |
XM_017017918.1 | 1142 | Missense Mutation | GGT,GTT | G,V 266 | XP_016873407.1 |