Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTGCTGGCATCCATGAAGCACAC[C/T]TCCTCTTCTTCATACTCTTCCTCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
OR6Q1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OR6Q1 - olfactory receptor family 6 subfamily Q member 1 (gene/pseudogene) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005186.2 | 79 | Missense Mutation | CTC,TTC | L,F 27 | NP_001005186.2 |
OR9Q1 - olfactory receptor family 9 subfamily Q member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005212.3 | 79 | Intron | NP_001005212.1 |
VN2R9P - vomeronasal 2 receptor 9 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |