Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGAAAGCATTGTGTCCTCAGATTGA[C/T]ATCAGGGTATATGTGAGAAAGATAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614908 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HIKESHI PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HIKESHI - Hikeshi, heat shock protein nuclear import factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001322404.1 | Intron | NP_001309333.1 | ||||
NM_001322407.1 | Intron | NP_001309336.1 | ||||
NM_001322409.1 | Intron | NP_001309338.1 | ||||
NM_016401.3 | Intron | NP_057485.2 | ||||
XM_017017914.1 | Intron | XP_016873403.1 | ||||
XM_017017915.1 | Intron | XP_016873404.1 |