Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAATCCTAACTAACTGTTTTTGTAA[A/G]TATTTTATTGAAACATTGCCACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609766 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CWC15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CWC15 - CWC15 spliceosome-associated protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016403.3 | Intron | NP_057487.2 | ||||
XM_011542868.2 | Intron | XP_011541170.1 | ||||
XM_017017916.1 | Intron | XP_016873405.1 |
KDM4D - lysine demethylase 4D | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |