Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTATCTTTCATTCCATTGGCCAGA[C/T]GCGCCATTCCCCAAGTGAGTTCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615292 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM111A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM111A - family with sequence similarity 111 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142519.2 | 319 | UTR 5 | NP_001135991.1 | |||
NM_001142520.2 | 319 | UTR 5 | NP_001135992.1 | |||
NM_001142521.2 | 319 | Intron | NP_001135993.1 | |||
NM_001312909.1 | 319 | UTR 5 | NP_001299838.1 | |||
NM_001312910.1 | 319 | UTR 5 | NP_001299839.1 | |||
NM_001312911.1 | 319 | Intron | NP_001299840.1 | |||
NM_022074.3 | 319 | Intron | NP_071357.2 | |||
NM_198847.2 | 319 | Intron | NP_942144.1 |
LOC101927204 - uncharacterized LOC101927204 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |