Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCAAATAAATGTTTAATATTAAT[A/C]ATTCCCAAACTGACAAGAACACAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616110 MIM: 610390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTX4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DTX4 - deltex E3 ubiquitin ligase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300727.1 | 5339 | UTR 3 | NP_001287656.1 | |||
NM_015177.1 | 5339 | UTR 3 | NP_055992.1 | |||
XM_006718483.3 | 5339 | Intron | XP_006718546.1 |
MPEG1 - macrophage expressed 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039396.1 | 5339 | Intron | NP_001034485.1 |