Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAAAGGGCATATGGTGACCTTCT[A/G]TGCCCTAGAAACTCTTTTAGGTATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147138 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MS4A2 - membrane spanning 4-domains A2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000139.4 | Intron | NP_000130.1 | ||||
NM_001256916.1 | Intron | NP_001243845.1 | ||||
XM_005273846.4 | Intron | XP_005273903.1 | ||||
XM_011544850.2 | Intron | XP_011543152.1 | ||||
XM_017017362.1 | Intron | XP_016872851.1 |