Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCGGAACATTCATGCATGTTTTGGA[A/G]AGGGGAGAAGGCAGTTAACAAATAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MS4A15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MS4A15 - membrane spanning 4-domains A15 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098835.1 | Intron | NP_001092305.1 | ||||
NM_001278242.1 | Intron | NP_001265171.1 | ||||
NM_152717.2 | Intron | NP_689930.1 | ||||
XM_006718459.3 | Intron | XP_006718522.1 | ||||
XM_011544812.2 | Intron | XP_011543114.1 | ||||
XM_011544814.1 | Intron | XP_011543116.1 |