Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTTAGAGAAAGCTAGCTTGGGCCA[C/T]GGTACTGAGGGGAACCCCAATGAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606148 MIM: 600393 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FADS1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FADS1 - fatty acid desaturase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013402.4 | Intron | NP_037534.3 | ||||
XM_011545022.2 | Intron | XP_011543324.1 |
FEN1 - flap structure-specific endonuclease 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM258 - transmembrane protein 258 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |