Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACCCAAGTGGATGATGCTTGGAAA[A/G]AGTCAGGGTGGAAAGAGACAGGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606096 | ||||||||||||||||||||
Literature Links: |
LGALS12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LGALS12 - galectin 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142535.1 | Intron | NP_001136007.1 | ||||
NM_001142536.1 | Intron | NP_001136008.1 | ||||
NM_001142537.1 | Intron | NP_001136009.1 | ||||
NM_001142538.1 | Intron | NP_001136010.1 | ||||
NM_033101.3 | Intron | NP_149092.2 | ||||
XM_017018458.1 | Intron | XP_016873947.1 |