Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGTTCCTTTTCTTTTTGGAATCT[A/G]TTCCCTCCATTGCACCTGAAGGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607380 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRAF3IP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRAF3IP1 - TRAF3 interacting protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001139490.1 | Intron | NP_001132962.1 | ||||
NM_015650.3 | Intron | NP_056465.2 | ||||
XM_006712414.2 | Intron | XP_006712477.1 | ||||
XM_011510944.2 | Intron | XP_011509246.1 | ||||
XM_011510945.2 | Intron | XP_011509247.1 | ||||
XM_011510946.2 | Intron | XP_011509248.1 | ||||
XM_011510947.2 | Intron | XP_011509249.1 | ||||
XM_011510948.2 | Intron | XP_011509250.1 | ||||
XM_011510950.2 | Intron | XP_011509252.1 | ||||
XM_017003789.1 | Intron | XP_016859278.1 |