Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAGGATGAATACTGTAACCTC[C/T]AAGTCCAACCTGTCTTATAATACCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP6V1C2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP6V1C2 - ATPase H+ transporting V1 subunit C2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039362.1 | 190 | Silent Mutation | TCC,TCT | S,S 27 | NP_001034451.1 | |
NM_144583.3 | 190 | Silent Mutation | TCC,TCT | S,S 27 | NP_653184.2 | |
XM_011510339.2 | 190 | Silent Mutation | TCC,TCT | S,S 27 | XP_011508641.1 | |
XM_011510340.2 | 190 | Silent Mutation | TCC,TCT | S,S 27 | XP_011508642.1 | |
XM_011510341.2 | 190 | Silent Mutation | TCC,TCT | S,S 27 | XP_011508643.1 | |
XM_017003745.1 | 190 | Silent Mutation | TCC,TCT | S,S 27 | XP_016859234.1 |