Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACCATTAACAGGGATATAGTTCAG[A/G]CCTGCGAAACTTATTGTCAAAGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604002 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PQLC3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PQLC3 - PQ loop repeat containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ROCK2 - Rho associated coiled-coil containing protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321643.1 | Intron | NP_001308572.1 | ||||
NM_004850.4 | Intron | NP_004841.2 | ||||
XM_005246190.3 | Intron | XP_005246247.1 | ||||
XM_011510417.2 | Intron | XP_011508719.1 | ||||
XM_017005378.1 | Intron | XP_016860867.1 | ||||
XM_017005379.1 | Intron | XP_016860868.1 |