Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTTATTGCCAGTTCATTTGAGGT[A/G]GAAGATCTAGAAACACTATTTGTTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603093 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
B3GALT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
B3GALT1 - beta-1,3-galactosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020981.3 | Intron | NP_066191.1 | ||||
XM_005246931.3 | Intron | XP_005246988.1 | ||||
XM_006712819.3 | Intron | XP_006712882.1 | ||||
XM_011512085.2 | Intron | XP_011510387.1 |