Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTATATTTTGGTCCCTGAGAGTTA[C/T]AAAAAGGGAGCTGGACTCCAGTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606093 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC173 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC173 - coiled-coil domain containing 173 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001085447.1 | 1610 | Missense Mutation | TAT,TGT | Y,C 532 | NP_001078916.1 | |
XM_011510589.1 | 1610 | Missense Mutation | TAT,TGT | Y,C 508 | XP_011508891.1 | |
XM_011510590.1 | 1610 | Missense Mutation | TAT,TGT | Y,C 451 | XP_011508892.1 |
PPIG - peptidylprolyl isomerase G | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |