Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGCGGAGCCTGAGCTAACCAGGGA[A/C]CTTCACATCCACATGAAGACAGGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613240 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R1C PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R1C - protein phosphatase 1 regulatory inhibitor subunit 1C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080545.2 | Intron | NP_001074014.1 | ||||
NM_001261424.1 | Intron | NP_001248353.1 | ||||
NM_001261425.1 | Intron | NP_001248354.1 | ||||
XM_017003451.1 | Intron | XP_016858940.1 |