Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATGGGTACTCTTGAAGAAATAT[C/T]TTCAAGTCCTTGAACACTGGGGATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616495 MIM: 612941 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL6IP6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL6IP6 - ADP ribosylation factor like GTPase 6 interacting protein 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152522.5 | 349 | Intron | NP_689735.1 | |||
XM_006712326.3 | 349 | UTR 5 | XP_006712389.1 |
PRPF40A - pre-mRNA processing factor 40 homolog A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |