Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGATGGAGAATGTGGCTCTGTTGAA[C/T]CACTTCGGCTTAAAGAGATAACAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600558 | ||||||||||||||||||||
Literature Links: |
LOC105373805 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC105373805 - uncharacterized LOC105373805 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
STAT4 - signal transducer and activator of transcription 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243835.1 | 2435 | Missense Mutation | GAT,GGT | D,G 707 | NP_001230764.1 | |
NM_003151.3 | 2435 | Missense Mutation | GAT,GGT | D,G 707 | NP_003142.1 | |
XM_006712719.3 | 2435 | Missense Mutation | GAT,GGT | D,G 707 | XP_006712782.1 | |
XM_011511705.2 | 2435 | Missense Mutation | GAT,GGT | D,G 707 | XP_011510007.1 | |
XM_017004784.1 | 2435 | Intron | XP_016860273.1 |