Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTTCAGGGCTTATTTGCCGTAGTA[C/T]TCTGCAGGTATTCTGTGACATTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606537 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MYO1B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MYO1B - myosin IB | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005246572.1 | Intron | XP_005246629.1 | ||||
XM_017004156.1 | Intron | XP_016859645.1 | ||||
XM_017004157.1 | Intron | XP_016859646.1 | ||||
XM_017004158.1 | Intron | XP_016859647.1 |