Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTTATGATCACATCAGTCTTGCTA[C/T]CGCTGACTCGGGCACTGTGTTAATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604727 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
STK17B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STK17B - serine/threonine kinase 17b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004226.3 | 2441 | UTR 3 | NP_004217.1 | |||
XM_011512168.2 | 2441 | UTR 3 | XP_011510470.1 | |||
XM_011512169.1 | 2441 | UTR 3 | XP_011510471.1 | |||
XM_011512170.1 | 2441 | UTR 3 | XP_011510472.1 | |||
XM_011512171.2 | 2441 | UTR 3 | XP_011510473.1 |