Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCATAAGTTCCATAAGCTATCTT[C/T]ACTCCTTTTCATTTTTTTTTTCTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC150 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC150 - coiled-coil domain containing 150 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080539.1 | Intron | NP_001074008.1 | ||||
XM_006712438.3 | Intron | XP_006712501.1 | ||||
XM_006712439.3 | Intron | XP_006712502.1 | ||||
XM_011510985.2 | Intron | XP_011509287.1 | ||||
XM_011510987.2 | Intron | XP_011509289.1 | ||||
XM_011510988.2 | Intron | XP_011509290.1 | ||||
XM_017003861.1 | Intron | XP_016859350.1 | ||||
XM_017003862.1 | Intron | XP_016859351.1 | ||||
XM_017003863.1 | Intron | XP_016859352.1 | ||||
XM_017003864.1 | Intron | XP_016859353.1 | ||||
XM_017003865.1 | Intron | XP_016859354.1 | ||||
XM_017003866.1 | Intron | XP_016859355.1 |