Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAAGTTCATCATCTGAGTGTCCA[A/G]TAGCATCTGGGTTTGAAAGTGGATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
FAM117B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FAM117B - family with sequence similarity 117 member B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ICA1L - islet cell autoantigen 1 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001288622.1 | 1916 | Missense Mutation | ACT,ATT | T,I 472 | NP_001275551.1 | |
NM_001288623.1 | 1916 | Missense Mutation | ACT,ATT | T,I 472 | NP_001275552.1 | |
NM_001288624.1 | 1916 | Intron | NP_001275553.1 | |||
NM_138468.5 | 1916 | Missense Mutation | ACT,ATT | T,I 472 | NP_612477.3 |