Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCCGCAACGTTGCATAGCCTGAT[A/G]TATGACTGCCTCCGTAACCAGCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606442 MIM: 609035 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABI2 - abl-interactor 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RAPH1 - Ras association (RalGDS/AF-6) and pleckstrin homology domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_203365.3 | 3749 | Intron | NP_976241.1 | |||
NM_213589.2 | 3749 | Missense Mutation | ACA,ATA | T,I 1223 | NP_998754.1 | |
XM_005246777.3 | 3749 | Missense Mutation | ACA,ATA | T,I 1275 | XP_005246834.1 | |
XM_005246779.4 | 3749 | Missense Mutation | ACA,ATA | T,I 1275 | XP_005246836.1 | |
XM_006712695.3 | 3749 | Missense Mutation | ACA,ATA | T,I 1275 | XP_006712758.1 | |
XM_011511646.2 | 3749 | Missense Mutation | ACA,ATA | T,I 1275 | XP_011509948.1 | |
XM_011511647.1 | 3749 | Missense Mutation | ACA,ATA | T,I 1250 | XP_011509949.1 | |
XM_011511648.1 | 3749 | Missense Mutation | ACA,ATA | T,I 1248 | XP_011509950.1 | |
XM_011511649.2 | 3749 | Intron | XP_011509951.1 |