Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAATTTTCCACGAGAAAAGTGAG[A/G]TAAGACATTCTAGGTGGAGGGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PARD3B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PARD3B - par-3 family cell polarity regulator beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302769.1 | Intron | NP_001289698.1 | ||||
NM_057177.6 | Intron | NP_476518.4 | ||||
NM_152526.5 | Intron | NP_689739.4 | ||||
NM_205863.3 | Intron | NP_995585.2 | ||||
XM_011510552.2 | Intron | XP_011508854.1 | ||||
XM_011510553.2 | Intron | XP_011508855.1 | ||||
XM_017003283.1 | Intron | XP_016858772.1 | ||||
XM_017003284.1 | Intron | XP_016858773.1 | ||||
XM_017003285.1 | Intron | XP_016858774.1 | ||||
XM_017003286.1 | Intron | XP_016858775.1 | ||||
XM_017003287.1 | Intron | XP_016858776.1 | ||||
XM_017003288.1 | Intron | XP_016858777.1 | ||||
XM_017003289.1 | Intron | XP_016858778.1 | ||||
XM_017003290.1 | Intron | XP_016858779.1 | ||||
XM_017003291.1 | Intron | XP_016858780.1 | ||||
XM_017003292.1 | Intron | XP_016858781.1 | ||||
XM_017003293.1 | Intron | XP_016858782.1 | ||||
XM_017003294.1 | Intron | XP_016858783.1 |