Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTAAGAGTTTTATACTCATTAAG[C/T]AGGGGGAAGGGGAGAACAAAAAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
INO80D PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
INO80D - INO80 complex subunit D | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017759.4 | 4143 | UTR 3 | NP_060229.3 | |||
XM_011511369.2 | 4143 | UTR 3 | XP_011509671.1 | |||
XM_011511370.2 | 4143 | Intron | XP_011509672.1 | |||
XM_011511371.2 | 4143 | Intron | XP_011509673.1 | |||
XM_011511373.2 | 4143 | Intron | XP_011509675.1 | |||
XM_011511375.2 | 4143 | UTR 3 | XP_011509677.1 | |||
XM_011511376.2 | 4143 | UTR 3 | XP_011509678.1 |