Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAATTTTATACAAGACACAGGTATA[C/T]GGAAACACAAATAACTTTAATGATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601593 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BARD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BARD1 - BRCA1 associated RING domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000465.3 | 2981 | UTR 3 | NP_000456.2 | |||
NM_001282543.1 | 2981 | UTR 3 | NP_001269472.1 | |||
NM_001282545.1 | 2981 | UTR 3 | NP_001269474.1 | |||
NM_001282548.1 | 2981 | UTR 3 | NP_001269477.1 | |||
NM_001282549.1 | 2981 | UTR 3 | NP_001269478.1 | |||
XM_017004613.1 | 2981 | UTR 3 | XP_016860102.1 | |||
XM_017004614.1 | 2981 | Intron | XP_016860103.1 |