Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGCAGAGCACAAGCGGGAGATTTA[C/T]GACCGCTATGGCCGGGAAGGGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604139 MIM: 601773 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DNAJB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DNAJB2 - DnaJ heat shock protein family (Hsp40) member B2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039550.1 | 432 | Silent Mutation | TAC,TAT | Y,Y 65 | NP_001034639.1 | |
NM_006736.5 | 432 | Silent Mutation | TAC,TAT | Y,Y 65 | NP_006727.2 |
PTPRN - protein tyrosine phosphatase, receptor type N | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TUBA4B - tubulin alpha 4b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |