Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTCAGGTAATCTATTCAACTGGA[C/T]ACTTTAAGCTTCATCTTTTTTTAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138430 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPD2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPD2 - glycerol-3-phosphate dehydrogenase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000408.4 | Intron | NP_000399.3 | ||||
NM_001083112.2 | Intron | NP_001076581.2 | ||||
XM_005246469.2 | Intron | XP_005246526.1 | ||||
XM_011510977.2 | Intron | XP_011509279.1 | ||||
XM_011510978.2 | Intron | XP_011509280.1 | ||||
XM_011510980.2 | Intron | XP_011509282.1 | ||||
XM_017003830.1 | Intron | XP_016859319.1 | ||||
XM_017003831.1 | Intron | XP_016859320.1 |