Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGGTCCATTGGAATCCAGATTT[G/T]CAAAGCATGTCAAGTTATGGCAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606152 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC19A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC19A3 - solute carrier family 19 member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_025243.3 | 1760 | UTR 3 | NP_079519.1 | |||
XM_005246874.3 | 1760 | UTR 3 | XP_005246931.1 | |||
XM_011511931.2 | 1760 | UTR 3 | XP_011510233.1 | |||
XM_011511932.1 | 1760 | UTR 3 | XP_011510234.1 | |||
XM_011511933.1 | 1760 | UTR 3 | XP_011510235.1 | |||
XM_017005030.1 | 1760 | UTR 3 | XP_016860519.1 | |||
XM_017005031.1 | 1760 | UTR 3 | XP_016860520.1 | |||
XM_017005032.1 | 1760 | UTR 3 | XP_016860521.1 | |||
XM_017005033.1 | 1760 | UTR 3 | XP_016860522.1 | |||
XM_017005034.1 | 1760 | UTR 3 | XP_016860523.1 |