Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCCGTCAAATGACTCCTTTAATG[C/T]CTCTTCAGTGGTATCCTCAGACAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 164035 MIM: 604012 MIM: 611133 | ||||||||||||||||||||
Literature Links: |
NCL PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NCL - nucleolin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005381.2 | 1900 | Missense Mutation | ACA,GCA | T,A 587 | NP_005372.2 |
SNORA75 - small nucleolar RNA, H/ACA box 75 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD20 - small nucleolar RNA, C/D box 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNORD82 - small nucleolar RNA, C/D box 82 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |