Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTCTCATTGACATGTGTGGGTTC[A/G]GTTGACAGGCGGGTCGAGTCTGAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607505 MIM: 602877 | ||||||||||||||||||||
Literature Links: |
PASK PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PASK - PAS domain containing serine/threonine kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PPP1R7 - protein phosphatase 1 regulatory subunit 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282409.1 | Intron | NP_001269338.1 | ||||
NM_001282410.1 | Intron | NP_001269339.1 | ||||
NM_001282411.1 | Intron | NP_001269340.1 | ||||
NM_001282412.1 | Intron | NP_001269341.1 | ||||
NM_001282413.1 | Intron | NP_001269342.1 | ||||
NM_001282414.1 | Intron | NP_001269343.1 | ||||
NM_002712.2 | Intron | NP_002703.1 | ||||
XM_011511388.1 | Intron | XP_011509690.1 |