Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGCACCGTCTCCAGGGGCAAACC[G/T]CTGTCCACCCCTGGCGACGACCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LINC01237 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LINC01237 - long intergenic non-protein coding RNA 1237 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RTP5 - receptor transporter protein 5 (putative) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173821.2 | 584 | Silent Mutation | CCG,CCT | P,P 187 | NP_776182.2 | |
XM_011510995.2 | 584 | Silent Mutation | CCG,CCT | P,P 166 | XP_011509297.1 | |
XM_011510997.2 | 584 | Silent Mutation | CCG,CCT | P,P 187 | XP_011509299.1 |