Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCGGAGGATGCTCCGGGCAACTTC[A/G]ACATTATTCTGGGCTATCTCTAAGG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 604491 | |||||||||||||||||||||||
Literature Links: |
CBLB PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CBLB - Cbl proto-oncogene B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321786.1 | 2766 | Silent Mutation | GTC,GTT | V,V 988 | NP_001308715.1 | |
NM_001321788.1 | 2766 | Silent Mutation | GTC,GTT | V,V 960 | NP_001308717.1 | |
NM_001321789.1 | 2766 | Silent Mutation | GTC,GTT | V,V 939 | NP_001308718.1 | |
NM_001321790.1 | 2766 | Silent Mutation | GTC,GTT | V,V 938 | NP_001308719.1 | |
NM_001321791.1 | 2766 | Silent Mutation | GTC,GTT | V,V 916 | NP_001308720.1 | |
NM_001321793.1 | 2766 | Silent Mutation | GTC,GTT | V,V 916 | NP_001308722.1 | |
NM_001321794.1 | 2766 | Silent Mutation | GTC,GTT | V,V 911 | NP_001308723.1 | |
NM_001321795.1 | 2766 | Silent Mutation | GTC,GTT | V,V 911 | NP_001308724.1 | |
NM_001321796.1 | 2766 | Silent Mutation | GTC,GTT | V,V 911 | NP_001308725.1 | |
NM_001321797.1 | 2766 | Silent Mutation | GTC,GTT | V,V 867 | NP_001308726.1 | |
NM_001321798.1 | 2766 | Silent Mutation | GTC,GTT | V,V 867 | NP_001308727.1 | |
NM_001321799.1 | 2766 | Silent Mutation | GTC,GTT | V,V 867 | NP_001308728.1 | |
NM_001321806.1 | 2766 | Silent Mutation | GTC,GTT | V,V 700 | NP_001308735.1 | |
NM_001321807.1 | 2766 | Silent Mutation | GTC,GTT | V,V 700 | NP_001308736.1 | |
NM_001321808.1 | 2766 | Silent Mutation | GTC,GTT | V,V 656 | NP_001308737.1 | |
NM_001321811.1 | 2766 | Silent Mutation | GTC,GTT | V,V 651 | NP_001308740.1 | |
NM_001321813.1 | 2766 | Silent Mutation | GTC,GTT | V,V 651 | NP_001308742.1 | |
NM_001321816.1 | 2766 | Silent Mutation | GTC,GTT | V,V 607 | NP_001308745.1 | |
NM_001321820.1 | 2766 | Silent Mutation | GTC,GTT | V,V 520 | NP_001308749.1 | |
NM_001321822.1 | 2766 | Silent Mutation | GTC,GTT | V,V 517 | NP_001308751.1 | |
NM_170662.4 | 2766 | Silent Mutation | GTC,GTT | V,V 960 | NP_733762.2 | |
XM_011513257.1 | 2766 | Silent Mutation | GTC,GTT | V,V 960 | XP_011511559.1 | |
XM_011513259.2 | 2766 | Intron | XP_011511561.2 | |||
XM_017007395.1 | 2766 | Silent Mutation | GTC,GTT | V,V 982 | XP_016862884.1 | |
XM_017007396.1 | 2766 | Silent Mutation | GTC,GTT | V,V 944 | XP_016862885.1 | |
XM_017007397.1 | 2766 | Silent Mutation | GTC,GTT | V,V 939 | XP_016862886.1 | |
XM_017007398.1 | 2766 | Silent Mutation | GTC,GTT | V,V 933 | XP_016862887.1 | |
XM_017007399.1 | 2766 | Silent Mutation | GTC,GTT | V,V 911 | XP_016862888.1 | |
XM_017007400.1 | 2766 | Silent Mutation | GTC,GTT | V,V 895 | XP_016862889.1 |