Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAATCTTACTGCTTTGTTTTTATC[A/G]TGTGTCCCAGGGAAATCAGATGCAG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
BBX PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
BBX - BBX, HMG-box containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142568.2 | Intron | NP_001136040.1 | ||||
NM_001276286.1 | Intron | NP_001263215.1 | ||||
NM_020235.6 | Intron | NP_064620.2 | ||||
XM_005247642.3 | Intron | XP_005247699.1 | ||||
XM_005247643.3 | Intron | XP_005247700.1 | ||||
XM_005247644.3 | Intron | XP_005247701.1 | ||||
XM_011512999.2 | Intron | XP_011511301.1 | ||||
XM_011513000.1 | Intron | XP_011511302.1 | ||||
XM_011513001.1 | Intron | XP_011511303.1 | ||||
XM_011513002.2 | Intron | XP_011511304.1 | ||||
XM_011513003.2 | Intron | XP_011511305.1 | ||||
XM_011513004.2 | Intron | XP_011511306.1 | ||||
XM_011513005.2 | Intron | XP_011511307.1 | ||||
XM_011513006.2 | Intron | XP_011511308.1 | ||||
XM_011513007.2 | Intron | XP_011511309.1 | ||||
XM_011513010.2 | Intron | XP_011511312.1 | ||||
XM_017006881.1 | Intron | XP_016862370.1 | ||||
XM_017006882.1 | Intron | XP_016862371.1 | ||||
XM_017006883.1 | Intron | XP_016862372.1 |