Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAATAAATAACACAGTGAAGAGG[G/T]GGAAGCAAAACAGAATCATCTTTAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 601028 | |||||||||||||||||||||||
Literature Links: |
CD47 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CD47 - CD47 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001777.3 | Intron | NP_001768.1 | ||||
NM_198793.2 | Intron | NP_942088.1 | ||||
XM_005247908.1 | Intron | XP_005247965.1 | ||||
XM_005247909.1 | Intron | XP_005247966.1 | ||||
XM_017007536.1 | Intron | XP_016863025.1 |