Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCCAGCACATCCTTGTAGAACTT[A/G]TCATTGTAGCTGACTGGAAAGATGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 610834 | ||||||||||||||||||||
Literature Links: |
NAA50 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NAA50 - N(alpha)-acetyltransferase 50, NatE catalytic subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308445.1 | 446 | Silent Mutation | GAC,GAT | D,D 32 | NP_001295374.1 | |
NM_025146.3 | 446 | Silent Mutation | GAC,GAT | D,D 33 | NP_079422.1 |