Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACAAACTTCTTCCATGTCCGAAG[C/G]GTGATCCAGAGGATCCTCCTTTAAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
CCDC191 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCDC191 - coiled-coil domain containing 191 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020817.1 | 2714 | Silent Mutation | ACC,ACG | T,T 875 | NP_065868.1 | |
XM_005247681.3 | 2714 | Silent Mutation | ACC,ACG | T,T 554 | XP_005247738.1 | |
XM_006713715.1 | 2714 | Silent Mutation | ACC,ACG | T,T 862 | XP_006713778.1 | |
XM_011513051.1 | 2714 | Silent Mutation | ACC,ACG | T,T 833 | XP_011511353.1 | |
XM_011513052.2 | 2714 | Silent Mutation | ACC,ACG | T,T 813 | XP_011511354.1 | |
XM_011513053.1 | 2714 | Silent Mutation | ACC,ACG | T,T 790 | XP_011511355.1 | |
XM_011513054.2 | 2714 | Silent Mutation | ACC,ACG | T,T 790 | XP_011511356.1 | |
XM_017006956.1 | 2714 | Silent Mutation | ACC,ACG | T,T 853 | XP_016862445.1 | |
XM_017006957.1 | 2714 | Silent Mutation | ACC,ACG | T,T 790 | XP_016862446.1 | |
XM_017006958.1 | 2714 | Silent Mutation | ACC,ACG | T,T 790 | XP_016862447.1 | |
XM_017006959.1 | 2714 | Silent Mutation | ACC,ACG | T,T 475 | XP_016862448.1 |
ZDHHC23 - zinc finger DHHC-type containing 23 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320466.1 | 2714 | Intron | NP_001307395.1 | |||
NM_001320467.1 | 2714 | Intron | NP_001307396.1 | |||
NM_001320468.1 | 2714 | Intron | NP_001307397.1 | |||
NM_173570.4 | 2714 | Intron | NP_775841.2 | |||
XM_005247269.1 | 2714 | Intron | XP_005247326.1 | |||
XM_005247270.3 | 2714 | Intron | XP_005247327.1 | |||
XM_006713562.3 | 2714 | Intron | XP_006713625.1 | |||
XM_011512618.2 | 2714 | Intron | XP_011510920.1 | |||
XM_011512619.1 | 2714 | Intron | XP_011510921.1 | |||
XM_017006084.1 | 2714 | Intron | XP_016861573.1 | |||
XM_017006085.1 | 2714 | Intron | XP_016861574.1 | |||
XM_017006086.1 | 2714 | Intron | XP_016861575.1 | |||
XM_017006087.1 | 2714 | Intron | XP_016861576.1 |