Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGCGTCGCCGACACCTCTGAGATA[A/G]AAGGGCCCCTTTCGACTAGCCTCTG
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
CCDC191 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CCDC191 - coiled-coil domain containing 191 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020817.1 | 137 | Intron | NP_065868.1 | |||
XM_005247681.3 | 137 | Intron | XP_005247738.1 | |||
XM_006713715.1 | 137 | Intron | XP_006713778.1 | |||
XM_011513051.1 | 137 | Intron | XP_011511353.1 | |||
XM_011513052.2 | 137 | Intron | XP_011511354.1 | |||
XM_011513053.1 | 137 | Intron | XP_011511355.1 | |||
XM_011513054.2 | 137 | Intron | XP_011511356.1 | |||
XM_017006956.1 | 137 | Intron | XP_016862445.1 | |||
XM_017006957.1 | 137 | Intron | XP_016862446.1 | |||
XM_017006958.1 | 137 | Intron | XP_016862447.1 | |||
XM_017006959.1 | 137 | Intron | XP_016862448.1 |
QTRT2 - queuine tRNA-ribosyltransferase accessory subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256835.1 | 137 | Intron | NP_001243764.1 | |||
NM_001256836.1 | 137 | Intron | NP_001243765.1 | |||
NM_001256837.1 | 137 | Intron | NP_001243766.1 | |||
NM_024638.3 | 137 | UTR 5 | NP_078914.1 | |||
XM_005247769.3 | 137 | UTR 5 | XP_005247826.1 | |||
XM_005247770.3 | 137 | Intron | XP_005247827.1 | |||
XM_006713744.3 | 137 | Intron | XP_006713807.1 | |||
XM_011513137.2 | 137 | Intron | XP_011511439.1 | |||
XM_017007182.1 | 137 | UTR 5 | XP_016862671.1 | |||
XM_017007183.1 | 137 | UTR 5 | XP_016862672.1 | |||
XM_017007184.1 | 137 | UTR 5 | XP_016862673.1 |