Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGGAGTAAAAAGAAGATAGCTGG[A/G]AGTGGATACAGCAGACATATAGCCC
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 603241 | |||||||||||||||||||||||
Literature Links: |
LSAMP PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LSAMP - limbic system-associated membrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318915.1 | 6719 | UTR 3 | NP_001305844.1 | |||
NM_002338.4 | 6719 | UTR 3 | NP_002329.2 | |||
XM_011512840.2 | 6719 | Intron | XP_011511142.1 | |||
XM_017006383.1 | 6719 | Intron | XP_016861872.1 | |||
XM_017006384.1 | 6719 | Intron | XP_016861873.1 |