Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTTTTATTGTCGTTTAGTTTT[C/T]AATATTCTGTACTTTTCATTGTGAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 607460 | |||||||||||||||||||||||
Literature Links: |
PLA1A PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PLA1A - phospholipase A1 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206960.1 | Intron | NP_001193889.1 | ||||
NM_001206961.1 | Intron | NP_001193890.1 | ||||
NM_001293225.1 | Intron | NP_001280154.1 | ||||
NM_015900.3 | Intron | NP_056984.1 | ||||
XM_017006572.1 | Intron | XP_016862061.1 |